Learn More
RETSCH Stainless-Steel Test Sieves, 150 mm Dia., in Millimeter Pore Sizes with ASTM E 11
Product Code. 10400212 Shop All Retsch Products
Change view
Click to view available options
Pore Size:
1 mm
1.18 mm
1.4 mm
1.7 mm
2 mm
2.36 mm
2.8 mm
3.35 mm
4 mm
4.75 mm
5.6 mm
6.3 mm
6.7 mm
8 mm
9.5 mm
11.2 mm
12.5 mm
13.2 mm
16 mm
19 mm
22.4 mm
25 mm
26.5 mm
31.5 mm
37.5 mm
45 mm
Unit Size:
Each
Product Code. Pore Size unitSize
10400212 1.18 mm Each
10259161 1.4 mm Each
10156021 1.7 mm Each
10585252 11.2 mm Each
10116311 12.5 mm Each
10618751 13.2 mm Each
10186401 16 mm Each
10496801 19 mm Each
10014801 2.36 mm Each
10583522 1 mm Each
10584482 2.8 mm Each
10712003 22.4 mm Each
10004721 26.5 mm Each
10196401 25 mm Each
10044811 3.35 mm Each
10380712 31.5 mm Each
10544102 2 mm Each
10044821 4.75 mm Each
10146501 37.5 mm Each
10156501 45 mm Each
10379511 4 mm Each
10340522 6.3 mm Each
10196161 5.6 mm Each
10156211 6.7 mm Each
10258491 8 mm Each
10380472 9.5 mm Each
26 options
Product Code. 10400212
Brand: RETSCH 60.117.001180
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Quantity 1 Each
Pore Size 1.18 mm
Type Test Sieve
Certifications/Compliance ASTM E 11
For Use With (Equipment) Shaker AS 200
Includes Certificate indicating Compliance
Height (Metric) 40mm
For Use With (Application) For separation, fractioning, particle size determination
Diameter (Metric) Outer 150 mm
Frame Material Stainless Steel
Mesh Material Stainless Steel