Learn More
- Home
- Products
- Filters and Filtration
- Mechanical Sieves
- RETSCH Stainless-Steel Test Sieves, 150 mm Dia., in Micrometer Pore Sizes with ISO 3310/1
RETSCH Stainless-Steel Test Sieves, 150 mm Dia., in Micrometer Pore Sizes with ISO 3310/1
Product Code. 10320622 Shop All Retsch Products| Product Code. | Pore Size | unitSize |
|---|---|---|
| 10320622 | 20 μm | Each |
| 10669311 | 100 μm | Each |
| 10515062 | 106 μm | Each |
| 10741423 | 112 μm | Each |
| 10298581 | 140 μm | Each |
| 10669501 | 150 μm | Each |
| 10309611 | 180 μm | Each |
| 10514872 | 160 μm | Each |
| 10741613 | 125 μm | Each |
| 10379701 | 200 μm | Each |
| 10420032 | 250 μm | Each |
| 10410032 | 224 μm | Each |
| 10339751 | 212 μm | Each |
| 10525642 | 25 μm | Each |
| 10585632 | 315 μm | Each |
| 10749312 | 280 μm | Each |
| 10437001 | 32 μm | Each |
| 10639881 | 300 μm | Each |
| 10759122 | 36 μm | Each |
| 10595632 | 355 μm | Each |
| 10608941 | 38 μm | Each |
| 10208781 | 400 μm | Each |
| 10769122 | 40 μm | Each |
| 10420042 | 450 μm | Each |
| 10410042 | 425 μm | Each |
| 10769312 | 45 μm | Each |
| 10004811 | 50 μm | Each |
| 10084721 | 500 μm | Each |
| 10136211 | 53 μm | Each |
| 10710083 | 560 μm | Each |
| 10249111 | 56 μm | Each |
| 10720083 | 630 μm | Each |
| 10320182 | 63 μm | Each |
| 10369941 | 600 μm | Each |
| 10450042 | 710 μm | Each |
| 10430022 | 71 μm | Each |
| 10379461 | 75 μm | Each |
| 10690442 | 800 μm | Each |
| 10155971 | 900 μm | Each |
| 10218971 | 850 μm | Each |
| 10440022 | 90 μm | Each |
| 10688931 | 80 μm | Each |
Product Code. 10320622
Brand: RETSCH 60.116.000020
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Specifications
| Quantity | 1 Each |
| Pore Size | 20 μm |
| Type | Test Sieve |
| Certifications/Compliance | ISO 3310/1 |
| For Use With (Equipment) | Shaker AS 200 |
| Includes | Certificate indicating Compliance |
| Height (Metric) | 40mm |
| For Use With (Application) | For separation, fractioning, particle size determination |
| Diameter (Metric) Outer | 150 mm |
| Frame Material | Stainless Steel |
| Mesh Material | Stainless Steel |
| Show More Show Less |