Learn More
RETSCH Stainless-Steel Test Sieves, 150 mm Dia., in Micrometer Pore Sizes with ASTM E 11
Product Code. 10555252 Shop All Retsch Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Pore Size:
20 μm
25 μm
32 μm
38 μm
45 μm
53 μm
63 μm
75 μm
90 μm
106 μm
125 μm
150 μm
180 μm
212 μm
250 μm
300 μm
355 μm
425 μm
500 μm
600 μm
710 μm
850 μm
missing translation for 'unitSize'
Each
Product Code. Pore Size unitSize
10555252 20 μm Each
10186591 106 μm Each
10430042 125 μm Each
10094721 150 μm Each
10536022 180 μm Each
10298821 212 μm Each
10595822 25 μm Each
10730083 250 μm Each
10044751 300 μm Each
10349751 32 μm Each
10330042 355 μm Each
10430032 38 μm Each
10536212 45 μm Each
10064761 425 μm Each
10600452 500 μm Each
10248681 53 μm Each
10573332 600 μm Each
10470032 63 μm Each
10490202 710 μm Each
10759312 75 μm Each
10218781 90 μm Each
10610452 850 μm Each
22 missing translation for 'options'
Product Code. 10555252
missing translation for 'mfr': RETSCH 60.117.000020
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Quantity 1 Each
Pore Size 20 μm
Type Test Sieve
Certifications/Compliance ASTM E 11
For Use With (Equipment) Shaker AS 200
Includes Certificate indicating Compliance
Height (Metric) 40mm
For Use With (Application) For separation, fractioning, particle size determination
Diameter (Metric) Outer 150 mm
Frame Material Stainless Steel
Mesh Material Stainless Steel