Learn More
- Home
- Products
- Filters and Filtration
- Mechanical Sieves
- RETSCH Stainless-Steel Test Sieves, 100 mm Dia., in Millimeter Pore Sizes with ISO 3310/1
RETSCH Stainless-Steel Test Sieves, 100 mm Dia., in Millimeter Pore Sizes with ISO 3310/1
Product Code. 10258921 Shop All Retsch Products
Change view
Click to view available options
Pore Size:
1 mm
1.12 mm
1.18 mm
1.25 mm
1.4 mm
1.6 mm
1.7 mm
1.8 mm
2 mm
2.24 mm
2.36 mm
2.5 mm
2.8 mm
3.15 mm
3.35 mm
3.55 mm
4 mm
4.5 mm
4.75 mm
5 mm
5.6 mm
6.3 mm
6.7 mm
7.1 mm
8 mm
9 mm
9.5 mm
10 mm
11.2 mm
12.5 mm
13.2 mm
14 mm
16 mm
18 mm
19 mm
20 mm
22.4 mm
25 mm
26.5 mm
28 mm
31.5 mm
35.5 mm
37.5 mm
40 mm
45 mm
Unit Size:
Each
| Product Code. | Pore Size | unitSize |
|---|---|---|
| 10258921 | 1.8 mm | Each |
| 10779502 | 1.12 mm | Each |
| 10298771 | 1.18 mm | Each |
| 10556212 | 1.25 mm | Each |
| 10516022 | 1.4 mm | Each |
| 10359941 | 1.6 mm | Each |
| 10610072 | 1.7 mm | Each |
| 10751233 | 10 mm | Each |
| 10024871 | 11.2 mm | Each |
| 10146261 | 12.5 mm | Each |
| 10751423 | 13.2 mm | Each |
| 10515642 | 18 mm | Each |
| 10106361 | 14 mm | Each |
| 10310622 | 16 mm | Each |
| 10116361 | 19 mm | Each |
| 10480032 | 1 mm | Each |
| 10740273 | 2.24 mm | Each |
| 10359991 | 2.36 mm | Each |
| 10074771 | 2.5 mm | Each |
| 10480042 | 2.8 mm | Each |
| 10457381 | 22.4 mm | Each |
| 10668751 | 25 mm | Each |
| 10545832 | 20 mm | Each |
| 10360132 | 28 mm | Each |
| 10136501 | 26.5 mm | Each |
| 10470042 | 2 mm | Each |
| 10209021 | 3.15 mm | Each |
| 10583332 | 3.35 mm | Each |
| 10700853 | 3.55 mm | Each |
| 10126211 | 31.5 mm | Each |
| 10459851 | 35.5 mm | Each |
| 10412162 | 4.5 mm | Each |
| 10514102 | 37.5 mm | Each |
| 10279111 | 4.75 mm | Each |
| 10209211 | 40 mm | Each |
| 10780843 | 45 mm | Each |
| 10106121 | 5.6 mm | Each |
| 10513722 | 4 mm | Each |
| 10196111 | 5 mm | Each |
| 10564292 | 6.3 mm | Each |
| 10426041 | 6.7 mm | Each |
| 10310382 | 7.1 mm | Each |
| 10456421 | 8 mm | Each |
| 10186161 | 9.5 mm | Each |
| 10466421 | 9 mm | Each |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Specifications
Specifications
| Quantity | 1 Each |
| Pore Size | 1.8 mm |
| Type | Test Sieve |
| Certifications/Compliance | ISO 3310/1 |
| For Use With (Equipment) | Shaker AS 200 |
| Includes | Certificate indicating Compliance |
| Height (Metric) | 40mm |
| For Use With (Application) | For separation, fractioning, particle size determination |
| Diameter (Metric) Outer | 100 mm |
| Frame Material | Stainless Steel |
| Mesh Material | Stainless Steel |
| Show More Show Less |