Learn More
Memmert™ Stainless Steel Slide-In Drip Tray
Product Code. 17109634 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP260eco
HPP410eco
HPP750eco
IPP1060eco
IPP110eco
IPP260eco
IPP410eco
IPP750eco
UN110pa
Unit Size:
Each
For Use With (Application):
Do not use with Modified Chamber
Reinforced Chamber Option K1
Product Code. For Use With (Equipment) unitSize For Use With (Application)
17109634 HPP1060eco Each -
17110295 UN110pa Each Do not use with Modified Chamber
17129784 HPP260eco Each Do not use with Modified Chamber
17129874 HPP750eco Each Do not use with Modified Chamber
17140255 IPP750eco Each Reinforced Chamber Option K1
17150225 IPP410eco Each Do not use with Modified Chamber
17150265 IPP750eco Each Do not use with Modified Chamber
17159684 HPP110eco Each Do not use with Modified Chamber
17160185 IPP260eco Each -
17179824 HPP410eco Each Do not use with Modified Chamber
17189854 HPP750eco Each Reinforced Chamber Option K1
17190045 IPP1060eco Each -
17190095 IPP110eco Each Do not use with Modified Chamber
13 options
Product Code. 17109634
Brand: Memmert™ HPP1060ecoB32599
Only null left
Add to basket
Add to basket
Stainless steel slide-in bottom drip tray
  • May affect the temperature distribution
  • 15 mm rim
Specifications
Description Sliding drip tray with 15 mm rim
For Use With (Equipment) HPP1060eco
Type Drip Tray
Length (Metric) 15 mm rim