Learn More
Memmert™ Stainless Steel Slide-In Bottom Drip Tray for IF110, 15 mm
Product Code. 17566615 Shop All Memmert Products
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
Product Code. 17566615
Brand: Memmert™ IF110B02073
Only null left
Add to basket
Add to basket
Non-perforated slide-in drip trays constructed of stainless steel.
  • Accessory
  • May affect temperature distribution
  • Constructed of stainless steel
  • Rim height: 15 mm
Specifications
For Use With (Equipment) IF110 Incubator
Material Stainless Steel
Type Drip Tray
Height (Metric) Rim: 15 mm