Learn More
RETSCH Stainless-Steel Lid for Test Sieves
Product Code. 13464886 Shop All Retsch Products
Change view
Click to view available options
Diameter (Metric) Outer:
100 mm
150 mm
200 mm
305 mm
315 mm
Unit Size:
Each
Product Code. Diameter (Metric) Outer unitSize
13464886 200 mm Each
10781033 100 mm Each
10791033 150 mm Each
11998124 315 mm Each
10659311 305 mm Each
5 options
Product Code. 13464886
Brand: RETSCH 69.520.0051
Only null left
Add to basket
Add to basket
Cover sieves to avoid loss of sample during separation process. These stainless steel lids will fit multiple test sieves, based upon the sieve's diameter.
  • Stainless steel
  • Available for a variety of test sieves, based upon diameter
Specifications
Quantity 1 Ea.
Type Sieve Cover
Certifications/Compliance ISO 3310 and ASTM E 11
Cover Material Stainless Steel
Diameter (Metric) Outer 200 mm