Learn More
RETSCH Stainless Steel Sieve Insert For Cyclone Mill Twister
Shop All Retsch Products
Click to view available options
Pore Size:
1 mm
2 mm
500 μm
800 μm
Unit Size:
Each
Specifications
Specifications
| Quantity | 1Each |
| Pore Size | 1 mm |
| Type | Sieve Insert |
| Material | Stainless Steel |
| For Use With (Equipment) | Cyclone Mill Twister |