Learn More
Fisherbrand™ Stainless Steel Sharp Ends Forceps
Product Code. 11562113 Shop All Fisherbrand Products
Change view
Click to view available options
Height (Metric):
115 mm
130 mm
160 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) unitSize
11562113 115 mm Each
11572113 130 mm Each
11582113 160 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11562113
  • ONLINE EXCLUSIVE PRICE
    218.00 NOK
    179.15 NOK / Pack of 1
    Save 38.85 NOK
    17% Off
View all promotions
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
Provides a non-stick surface with PTFE coating to which very few solid or liquid substances will permanently adhere. Nickel Electro™ Stainless Steel Sharp Ends Forceps provide excellent non-wetting properties for easy cleaning and can be steam or chemically sterilized.
PTFE coated forceps, providing a non-stick surface to which very few solid or liquid substances will permanently adhere.
  • Chemically resistant and non-contaminating
  • Non-protein binding and providing bio-inert surfaces
  • No critical quantities lost due to sticking
  • No build up on surfaces
  • Ideal for use under severe laboratory conditions
  • Excellent non-wetting properties for easy cleaning and can be steam or chemically sterilized
  • With guide pin
Specifications
Product Type Forceps
Material PTFE-coated Stainless Steel
Height (Metric) 115 mm
No. per Pack Each
Style Serrated Jaws
Tip Style Sharp