Specifications
| Quantity | 1 Each |
| Pore Size | 250 μm |
| Type | Test Sieve |
| Certifications/Compliance | ISO 3310 and ASTM E 11 |
| Height (Metric) | 25mm |
| For Use With (Application) | Separation, fractioning, particle size determination |
| Diameter (Metric) Outer | 200 mm |
| Frame Material | Stainless Steel |
| Mesh Material | Stainless Steel |