Learn More
Fisherbrand™ Stainless Steel Chattaway Spatulas
Product Code. 11513472 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Length (Metric):
150 mm
178 mm
200 mm
Unit Size:
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
11513472 150 mm Pack of 5
11523472 178 mm Pack of 5
11573482 200 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11513472
  • ONLINE EXCLUSIVE PRICE
    254.00 NOK
    214.09 NOK / Pack of 5
    Save 39.91 NOK
    15% Off
Estimated Shipment: 08-01-2027
Add to basket
Add to basket
Scrapes and lifts materials from bottles or flasks. Fisherbrand™ Stainless Steel Chattaway Spatulas are non-magnetic with a bright finish.
  • Manufactured from Stainless Steel
  • Bent and rounded end scrapes and lifts materials from bottles and flasks
Specifications
Quantity 5 Pk.
Product Type Chattaway Spatula
Width (Metric) 10 mm
Diameter (Metric) Outer 3.6 mm
Dimensions (L x W) 150 x 10 mm
Material Stainless Steel
Length (Metric) 150 mm