Learn More
Fisherbrand™ Stainless Steel Manual Decapper for clean room
Product Code. 11845722 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Unit Size:
Each
Size:
13 mm
20 mm
32 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size Size
11845722 Each -
11520535 Each 20 mm
15259968 Each 32 mm
11586034 Each 13 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11845722
Quantity Price
1 9309.54 NOK / Pack of 1
4 8620.63 NOK / Pack of 1
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Allow easy removal of aluminum seals without breakage
  • Stainless Steel
  • Sterilizable for cleanroom applications
  • Manual
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Description 11mm Decapper
For Use With (Equipment) Aluminum Cap