Learn More
Fisherbrand™ Stainless Steel Manual Crimper for clean room, adjustable
Product Code. 11737746 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Diameter (Metric) Cap:
11 mm
13 mm
20 mm
32 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Cap Unit Size
11737746 11 mm Each
11757746 20 mm Each
11747746 13 mm Each
12686835 32 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11737746
Estimated Shipment: 23-11-2026
Add to basket
Add to basket
Provides a reproducible, secure vial closure
  • Stainless Steel
  • Sterilizable for cleanroom applications
  • Manual
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Description 11mm Crimper
Diameter (Metric) Cap 11 mm
Material Stainless Steel
Product Type Manual Crimper