Learn More
Bandelin Electronic™ Sonorex™ Insert Baskets for Ultrasonic Baths
Description
Facilitates ultrasonic applications and simultaneously protects oscillating tank and parts to be cleaned. Bandelin Electronic™ Sonorex™ Insert Baskets for Ultrasonic Baths are manufactured from stainless steel.
- Available for a wide range of tank capacities
- Available in a range of mesh sizes
- Excellent for the cleaning of technical glassware, analysis sieves, medical instruments, metal parts, electronic components, degassing, emulsifying and sample preparation
Specifications
Specifications
| Type | Basket Insert |
| For Use With (Equipment) | Ultrasonic Bath, RK 514 BH, DK 514 BP |
| Height (Metric) | 275 mm |
| Length (Metric) | 245 mm |
| Material | Stainless Steel |
| Width (Metric) | 50 mm |
| Dimensions (L x W x H) | 245 x 50 x 275 mm |
| Model | K14B |