Learn More
Socorex™ Stainless Steel Injection Needles
Product Code. 11807293
Change view
Click to view available options
Needle Gauge:
14 G
16 G
18 G
19 G
21 G
25 G
Length (Metric):
3 mm
25 mm
30 mm
40 mm
50 mm
Unit Size:
Pack of 12
Product Code. Needle Gauge Length (Metric) unitSize
11807293 14 G 50 mm Pack of 12
11857263 25 G 3 mm Pack of 12
11807273 21 G 25 mm Pack of 12
10594235 21 G 40 mm Pack of 12
11847273 19 G 30 mm Pack of 12
11867283 16 G 30 mm Pack of 12
11897283 14 G 40 mm Pack of 12
11867273 18 G 50 mm Pack of 12
8 options
Product Code. 11807293
Brand: Socorex™ 371.2050
Manufactured with Swiss made quality, Socorex™ Stainless Steel Injection Needles are Luer Lock needles that are reliable and durable. They include a dispensing box for easy and safe removal and dispensing of needles.
  • Stainless steel
  • Sharp bevel
  • Reusable
  • Autoclavable
  • Includes dispensing box with sliding lid for easy and safe needle removal
Specifications
Autoclavable Autoclavable
Type Injection Needle
Material Stainless Steel
Needle Gauge 14 G
Tip Style Sharp Bevel
Length (Metric) 50 mm
Diameter (Metric) 2 mm