Learn More
Socorex™ Stainless Steel Injection Needles
Description
- Stainless steel
- Sharp bevel
- Reusable
- Autoclavable
- Includes dispensing box with sliding lid for easy and safe needle removal
Specifications
Specifications
| Autoclavable | Autoclavable |
| Type | Injection Needle |
| Material | Stainless Steel |
| Needle Gauge | 21 G |
| Tip Style | Sharp Bevel |
| Length (Metric) | 25 mm |
| Diameter (Metric) | 0.8 mm |