Learn More
Fisherbrand™ Stainless Steel Cylindrical Calibration Weight Set, Class F2
Product Code. 15525129 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Weight (Metric):
1 g to 1 kg
1 g to 5 kg
1 to 100 g
Unit Size:
1 unit
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Weight (Metric) Unit Size
15525129 1 g to 1 kg 1 unit
15525119 1 to 100 g 1 unit
15505139 1 g to 5 kg 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15525129
  • ONLINE EXCLUSIVE PRICE
    9470.00 NOK
    6240.00 NOK / Pack of 1
    Save 3230.00 NOK
    34% Off
Estimated Shipment: 18-12-2026
Add to basket
Add to basket
Use with weighing instruments of high accuracy class II. Fisherbrand™ Class F2 Stainless Steel Calibration Weights are also intended for use in important commercial transactions, such as those involving precious metals and stones.
  • Class F2 stainless steel calibration weight
  • Weights intended for use in the verification or calibration of class M1 and possibly class M2 weights
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
Specifications
Classification F2
Weight (Metric) 1 g to 1 kg
Mass 1 g to 1 kg
Shape Cylindrical