Learn More
Fisherbrand™ Stainless Steel Cylindrical Calibration Weight Set, Class F2
Shop All Fisherbrand Products
Click to view available options
Weight (Metric):
1 g to 1 kg
1 g to 5 kg
1 to 100 g
Unit Size:
1 unit
Specifications
Specifications
| Classification | F2 |
| Weight (Metric) | 1 to 100 g |
| Mass | 1 g to 100 g |
| Shape | Cylindrical |