Learn More
RETSCH Stainless Steel Bottom Sieve For SK100 Cross Beater Mills
Shop All Retsch Products
Click to view available options
Pore Size:
1 mm
120 μm
2 mm
200 μm
250 μm
5 mm
500 μm
Unit Size:
Each
Specifications
Specifications
| Quantity | 1Each |
| Pore Size | 2 mm |
| Type | Bottom Sieve |
| Material | Stainless Steel |
| For Use With (Equipment) | SK100 Cross Beater Mills |