Learn More
RETSCH Stainless Steel Bottom Sieve For SK100 Cross Beater Mills
Product Code. 10489872 Shop All Retsch Products
Change view
Click to view available options
Pore Size:
1 mm
120 μm
2 mm
200 μm
250 μm
5 mm
500 μm
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size unitSize
10489872 500 μm Each
10410263 120 μm Each
10709171 250 μm Each
10039400 200 μm Each
10605121 1 mm Each
10759551 2 mm Each
10450453 5 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 10489872 Supplier RETSCH Supplier No. 02.407.0002
Only null left
Specifications
Quantity 1Each
Pore Size 500 μm
Type Bottom Sieve
Material Stainless Steel
For Use With (Equipment) SK100 Cross Beater Mills