Learn More
RETSCH Stainless Steel Bottom Sieve For Cutting Mill SM 100 and Heavy-Duty Cutting Mill SM 2000
Shop All Retsch Products
Click to view available options
Pore Size:
1 mm
500 μm
750 μm
Unit Size:
Each
Specifications
Specifications
| Quantity | 1Each |
| Pore Size | 500 μm |
| Type | Bottom Sieve |
| Material | Stainless Steel |
| For Use With (Equipment) | Cutting Mill SM 100 and Heavy-Duty Cutting Mill SM 2000 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion