Learn More
Memmert™ Stainless Steel Bottom Drip Tray for SF450plus, 15 mm
Product Code. 17588275 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
SF450plus Sterilizer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
17588275 SF450plus Sterilizer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17588275 Supplier Memmert™ Supplier No. SF450plusB04362
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
Non-perforated bottom drip trays constructed of stainless steel.
  • Accessory
  • May affect temperature distribution
  • Constructed of stainless steel
  • Rim height: 15 mm
Specifications
For Use With (Equipment) SF450plus Sterilizer
Product Type Bottom Drip Tray
Height (Metric) Tray Rim: 15 mm
Material Stainless Steel