Learn More
Memmert™ Stainless Steel Bottom Drip Tray
Product Code. 17179624 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
HPP1060eco, Not Option K1
HPP110eco, Not Option k1
HPP260eco
HPP410eco
HPP750eco, Not Option K1
HPP750eco, only with Option K1
IPP1060eco, Not Option K1
IPP110eco, Not Option K1
IPP260eco
IPP410eco
IPP750eco, Not Option K1
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
17179624 HPP1060eco, Not Option K1 Each
17109764 HPP260eco Each
17109864 HPP750eco, only with Option K1 Each
17119854 HPP750eco, Not Option K1 Each
17139664 HPP110eco, Not Option k1 Each
17160045 IPP1060eco, Not Option K1 Each
17160165 IPP260eco Each
17170245 IPP750eco, Not Option K1 Each
17180205 IPP410eco Each
17189804 HPP410eco Each
17190075 IPP110eco, Not Option K1 Each
11 options
Product Code. 17179624
Brand: Memmert™ HPP1060ecoB29769
Only null left
Add to basket
Add to basket
15 mm stainless steel bottom drip tray
  • May affect the temperature distribution
  • 15 mm
Specifications
Description Not in connection with option K1
For Use With (Equipment) HPP1060eco, Not Option K1
Diameter (English) 15 mm
Diameter (Metric) 15 mm