Learn More
ST6GALNAC6 Mouse anti-Human, Mouse, Clone: 1A7B7, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66209-1-IG-20UL
 View more versions of this product
Product Code. 16847875
  • 1710.33 NOK / 20µL
Description
ST6GALNAC6 belongs to a family of sialyltransferases that modify proteins and ceramides on the cell surface to alter cell-cell or cell-extracellular matrix interactions.
Specifications
Specifications
ST6GALNAC6
Monoclonal
0.74 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q969X2, Q9JM95
St6galnac6
ST6GALNAC6 Fusion Protein Ag8358
20 μL
Primary
Mouse, Human
Antibody
IgG1
Western Blot, Immunohistochemistry (Paraffin)
1A7B7
Unconjugated
ST6GALNAC6
hST6GalNAc VI, RP11 203J24.3, Sialyltransferase 7F, SIAT7 F, SIAT7F, ST6GalNAc VI, ST6GALNAC6, ST6GALNACVI
Mouse
Protein G
RUO
30815, 50935
-20°C