missing translation for 'onlineSavingsMsg'
Learn More

ST6GALNAC6 Mouse anti-Human, Mouse, Clone: 1A7B7, Proteintech

Product Code. p-7213118 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16847875 20 μL 20µL
16837875 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16847875

Brand: Proteintech 662091IG20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ST6GALNAC6 belongs to a family of sialyltransferases that modify proteins and ceramides on the cell surface to alter cell-cell or cell-extracellular matrix interactions.

Specifications

Antigen ST6GALNAC6
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1A7B7
Concentration 0.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ST6GALNAC6
Gene Accession No. Q969X2, Q9JM95
Gene Alias hST6GalNAc VI, RP11 203J24.3, Sialyltransferase 7F, SIAT7 F, SIAT7F, ST6GalNAc VI, ST6GALNAC6, ST6GALNACVI
Gene Symbols St6galnac6
Host Species Mouse
Immunogen ST6GALNAC6 Fusion Protein Ag8358
Purification Method Protein G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30815, 50935
Target Species Mouse, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.