Learn More
Spectrum™ Spectra/Por™ Float-A-Lyzer™ G2 Dialysis Devices
Product Code. p-2410196
Change view
Click to view available options
MWCO:
0.1 to 0.5 kDa
0.5 to 1 kDa
100 kDa
1000 kDa
20 kDa
3.5 to 5 kDa
300 kDa
50 kDa
8 to 10 kDa
Color Code:
Amber
Black
Blue
Green
Orange
Pink
Red
Violet
Yellow
Length (Metric):
50 mm
100 mm
160 mm
Unit Size:
Pack of 12
28 product options available for selection
Product selection table with 28 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. MWCO Color Code Length (Metric) Unit Size
11571160 100 kDa Blue 50 mm Pack of 12
11531160 3.5 to 5 kDa Black 50 mm Pack of 12
11541180 50 kDa Violet 160 mm Pack of 12
11561170 100 kDa Blue 100 mm Pack of 12
11581160 300 kDa Amber 50 mm Pack of 12
11511160 0.1 to 0.5 kDa Green 50 mm Pack of 12
11541160 8 to 10 kDa Yellow 50 mm Pack of 12
11551160 20 kDa Red 50 mm Pack of 12
11581170 0.1 to 0.5 kDa Green 160 mm Pack of 12
11591170 1000 kDa Pink 100 mm Pack of 12
11561160 50 kDa Violet 50 mm Pack of 12
11495829 8 to 10 kDa Yellow 160 mm Pack of 12
11521180 8 to 10 kDa Yellow 160 mm Pack of 12
11571180 1000 kDa Pink 160 mm Pack of 12
11501170 0.1 to 0.5 kDa Green 100 mm Pack of 12
11501180 0.5 to 1 kDa Orange 160 mm Pack of 12
11571170 300 kDa Amber 100 mm Pack of 12
11551170 50 kDa Violet 100 mm Pack of 12
11511170 0.5 to 1 kDa Orange 100 mm Pack of 12
11591160 1000 kDa Pink 50 mm Pack of 12
11561180 300 kDa Amber 160 mm Pack of 12
11521160 0.5 to 1 kDa Orange 50 mm Pack of 12
11541170 20 kDa Red 100 mm Pack of 12
11521170 3.5 to 5 kDa Black 100 mm Pack of 12
11531180 20 kDa Red 160 mm Pack of 12
11837271 8 to 10 kDa Yellow 100 mm Pack of 12
11827271 3.5 to 5 kDa Black 160 mm Pack of 12
11551180 100 kDa Blue 160 mm Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 28 options available.
28 options
Product Code. 11571160 Supplier Spectrum™ Supplier No. G235035
Ready-to-use and disposable, Float-A-Lyzer G2 incorporates biotech-grade cellulose ester (CE) membrane tubing into a preasembled dialysis device that is easy-to-use and provides ultimate convenience for small volume dialysis (1 to 10mL)
Available in volume sizes: 1mL, 5mL and 10 mL. Resealable screw cap is color-coded for 9 available MWCOs and provides open access for easy loading and total sample recovery with a disposable pipet (included for 5mL and 10mL sizes). Flotation ring (included) provides buoyancy to maximize dialysis efficiency.
  • Superior handling and leak protection
  • Resealable screw cap and pipet for easy loading and total recovery
  • Highest membrane purity, no heavy metals and sulfides
  • Minimizes volume specific dilution
  • 9 MWCOs ranging from 100 to 1,000,000 Daltons
Specifications
Product Type Float-A-Lyzer G2 Dialysis Device
MWCO 100 kDa
Membrane Cellulose Ester
Diameter (English) Membrane 0.39 in.
Diameter (English)Top 0.9 in.
Diameter (Metric) Membrane 10 mm
Diameter (Metric)Top 23 mm
Length (English) 1.9 in.
Length (Metric) 50 mm
Closure Material Polypropylene
Closure Type Screw Cap
Color Code Blue
Kit Contents Disposable pipette for 5 and 10mL, flotation ring
Quantity 12 Pack
For Use With (Application) Ready to use laboratory dialysis devices, highest membrane purity, superior handling and leak prevention, 95 to 98% sample recovery, perfect for dialyzing electro-eluted proteins or chromatography fractions