Learn More
Spectrum Labs™ Spectra/Por™ 2 Membrane Disc
Description
For use with Spectrum Equilibrium Dialyzer and Spectra/Por™ MacroDialyzer systems.
Regenerated cellulose (RC) precut membrane discs
Supplied dry with a glycerol as a humectant
Regenerated cellulose (RC) precut membrane discs
Supplied dry with a glycerol as a humectant
Specifications
Specifications
| Product Type | Membrane Disc |
| MWCO | 12 to 14 kDa |
| Membrane | Regenerated Cellulose |
| Diameter (Metric) | 100 mm |
| For Use With (Application) | For Higher Permeability |