Learn More
Spectrum Labs™ Spectra/Por™ 1 Membrane Disc
Product Code. 12707526
Change view
Click to view available options
Diameter (Metric):
47 mm
100 mm
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
12707526 47 mm Pack of 50
12717526 100 mm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12707526 Supplier Spectrum Labs™ Supplier No. 132476
Regenerated cellulose membrance for use with Spectrum Equilibrium Dializer and Spectra/Por MacroDializer systems
For use with Spectrum Equilibrium Dialyzer and Spectra/Por™ MacroDialyzer systems.
Regenerated cellulose (RC) precut membrane discs
Supplied dry with a glycerol as a humectant
Specifications
Product Type Membrane Disc
MWCO 6 to 8 kDa
Membrane Regenerated Cellulose
Diameter (Metric) 47 mm
For Use With (Application) For Basic Dialysis