Learn More
Gilson™ SPE Manifold
Product Code. 16696165 Shop All Gilson Products
Change view
Click to view available options
No. of Ports:
24
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Ports Unit Size
16696165 24 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16696165 Supplier Gilson™ Supplier No. 37417012
  • ONLINE EXCLUSIVE PRICE
    20170.00 NOK
    19108.51 NOK / Pack of 1
    Save 1061.49 NOK
    5% Off
 Restricted Item
Estimated Shipment: 10-11-2026
SPE Manifold Kit
  • Cartridge ejection plate (quantity 1)
  • Cartridge rack, SPE, 24-positions, 3 mL (quantity 1)
  • Gasket with adhesive-backed seal, 24-positions (quantity 2)
  • Manifold, SPE, 24-positions (quantity 1)
  • O-ring manifold seal (quantity 1)
  • Tube rack, SPE collection, 1–3 mL, 12 x 75 mm (quantity 1)
Specifications
Description 24 Position Kit
Volume (Metric) 3 mL
Material SPE Manifold
No. of Ports 24