Learn More
Eppendorf™ Spare Sealing
Product Code. 13256609 Shop All Eppendorf Products
Change view
Click to view available options
For Use With (Equipment):
750mL Bucket /S-4-104
Aerosol Tight Cap 400mL
FA-45-48-11
QuickLock Rotor Lid FA-45-48-11
Rotor A-2-DWP-AT
Rotor Lid S-24-11-AT
Rotor S-24-11-AT
Rotor lid
Unit Size:
Each
Pack of 2
Pack of 4
Pack of 5
Product Code. For Use With (Equipment) unitSize
13256609 750mL Bucket /S-4-104 Pack of 5
15100017 QuickLock Rotor Lid FA-45-48-11 Pack of 5
10580003 Aerosol Tight Cap 400mL Pack of 4
13286609 FA-45-48-11 Pack of 5
12803222 Rotor A-2-DWP-AT Pack of 2
15224546 Rotor S-24-11-AT Each
13206619 Rotor lid Pack of 5
15214546 Rotor Lid S-24-11-AT Pack of 5
8 options
Product Code. 13256609
Brand: Eppendorf™ 5820749008
For use with Eppendorf Rotors
  • Spare sealing for rotor lids
  • Available for a wide range of products
TRUSTED_SUSTAINABILITY
Specifications
Type Spare Sealing
For Use With (Equipment) 750mL Bucket /S-4-104
Includes Pack of 5