Learn More
Fisherbrand™ Spare Cartridges for Portable Demineralizers
Product Code. 11944169 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
For Use With (Equipment):
DEM'O3 Portable Demineralizer
Portable Demineralizer DEM'O4
Portable Demineralizer DEM'O5
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11944169 DEM'O3 Portable Demineralizer Each
11954169 Portable Demineralizer DEM'O4 Each
11964169 Portable Demineralizer DEM'O5 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11944169
  • ONLINE EXCLUSIVE PRICE
    2500.00 NOK
    2359.66 NOK / Pack of 1
    Save 140.34 NOK
    5% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
Facilitates the demineralization of water with conductivity of 0.3 to 2μS/cm and pH 6 to 7. Fisherbrand™ Spare Cartridges for Portable Demineralizers start blue and gradually change color to ocher as they become saturated.
  • Contain only new resins that have not been polluted or regenerated
  • Level of performance is constant and guaranteed
  • Robust and practical construction
  • Quick connectors make assembly/dismantling easy and fast
Specifications
Color Indicator Blue cartridge turns ocher brown
Capacity Depends on the quality of the inlet water
Flow Rate 150 L/hr.
Resin Type Ion Exchanger
Filter Size 60 cm
For Use With (Equipment) DEM'O3 Portable Demineralizer
Quantity 3/Set
Type Spare Cartridge