Learn More
Fisherbrand™ Solid Neoprene Rubber Bungs
Product Code. 11567113 Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (Metric) Bottom:
3 mm
4 mm
6 mm
7 mm
9 mm
10 mm
11 mm
13 mm
17 mm
19 mm
25 mm
27 mm
33 mm
39 mm
43 mm
49 mm
53 mm
57 mm
62 mm
67 mm
71 mm
75 mm
79 mm
86 mm
92 mm
99 mm
Diameter (Metric) Top:
4.5 mm
5.8 mm
8 mm
9 mm
11.5 mm
12.5 mm
14 mm
16 mm
20.3 mm
22.5 mm
28.5 mm
31 mm
38.5 mm
44.5 mm
49 mm
55 mm
59.5 mm
64 mm
69 mm
74 mm
78 mm
85 mm
89 mm
96 mm
102 mm
109 mm
Length (Metric):
12 mm
14 mm
16 mm
20 mm
24 mm
26 mm
28 mm
32 mm
38.5 mm
42 mm
45.5 mm
49 mm
54 mm
Unit Size:
Each
Pack of 10
Pack of 2
Pack of 20
Pack of 5
Pack of 50
Product Code. Diameter (Metric) Bottom Diameter (Metric) Top Length (Metric) unitSize
11567113 62 mm 69 mm 49 mm Each
11547093 10 mm 12.5 mm 20 mm Pack of 20
11587093 17 mm 20.3 mm 26 mm Pack of 20
11557093 11 mm 14 mm 24 mm Pack of 20
11597093 19 mm 22.5 mm 28 mm Pack of 10
11527103 25 mm 28.5 mm 28 mm Pack of 10
11537103 27 mm 31 mm 32 mm Pack of 10
11567103 33 mm 38.5 mm 38.5 mm Pack of 5
11597103 39 mm 44.5 mm 38.5 mm Pack of 5
11517113 43 mm 49 mm 42 mm Pack of 2
11537113 49 mm 55 mm 42 mm Pack of 2
11587083 4 mm 5.8 mm 14 mm Pack of 50
11547113 53 mm 59.5 mm 45.5 mm Each
11557113 57 mm 64 mm 49 mm Each
11577113 67 mm 74 mm 49 mm Each
11507093 6 mm 8 mm 16 mm Pack of 20
11587113 71 mm 78 mm 49 mm Each
11597113 75 mm 85 mm 54 mm Each
11517093 7 mm 9 mm 16 mm Pack of 20
11537093 9 mm 11.5 mm 20 mm Pack of 20
11507123 79 mm 89 mm 54 mm Each
11517123 86 mm 96 mm 54 mm Each
11527123 92 mm 102 mm 54 mm Each
11537123 99 mm 109 mm 54 mm Each
11577083 3 mm 4.5 mm 12 mm Pack of 50
10587064 3 mm 4.5 mm 12 mm Pack of 50
11567093 13 mm 16 mm 24 mm Pack of 20
10265114 13 mm 16 mm 24 mm Pack of 20
28 options
Product Code. 11567113
Used for laboratory purposes.
  • Made up of neoprene rubber
  • Hardness is 46/55 IRHD
Specifications
For Use With (Application) For laboratory use
Hardness 46/55 IRHD
Length (Metric) 49 mm
Quantity 1/Ea.
Color Gray
Diameter (Metric) Bottom 62 mm
Diameter (Metric) Top 69 mm
Material Neoprene
Product Type Bung