Learn More
- Home
- Products
- Stoppers and Closures
- Stoppers
- Fisherbrand™ Solid Neoprene Rubber Bungs
Fisherbrand™ Solid Neoprene Rubber Bungs
Product Code. 11547113 Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (Metric) Bottom:
3 mm
4 mm
6 mm
7 mm
9 mm
10 mm
11 mm
13 mm
17 mm
19 mm
25 mm
27 mm
33 mm
39 mm
43 mm
49 mm
53 mm
57 mm
62 mm
67 mm
71 mm
75 mm
79 mm
86 mm
92 mm
99 mm
Diameter (Metric) Top:
4.5 mm
5.8 mm
8 mm
9 mm
11.5 mm
12.5 mm
14 mm
16 mm
20.3 mm
22.5 mm
28.5 mm
31 mm
38.5 mm
44.5 mm
49 mm
55 mm
59.5 mm
64 mm
69 mm
74 mm
78 mm
85 mm
89 mm
96 mm
102 mm
109 mm
Length (Metric):
12 mm
14 mm
16 mm
20 mm
24 mm
26 mm
28 mm
32 mm
38.5 mm
42 mm
45.5 mm
49 mm
54 mm
Unit Size:
Each
Pack of 10
Pack of 2
Pack of 20
Pack of 5
Pack of 50
| Product Code. | Diameter (Metric) Bottom | Diameter (Metric) Top | Length (Metric) | unitSize |
|---|---|---|---|---|
| 11547113 | 53 mm | 59.5 mm | 45.5 mm | Each |
| 11547093 | 10 mm | 12.5 mm | 20 mm | Pack of 20 |
| 11587093 | 17 mm | 20.3 mm | 26 mm | Pack of 20 |
| 11557093 | 11 mm | 14 mm | 24 mm | Pack of 20 |
| 11597093 | 19 mm | 22.5 mm | 28 mm | Pack of 10 |
| 11527103 | 25 mm | 28.5 mm | 28 mm | Pack of 10 |
| 11537103 | 27 mm | 31 mm | 32 mm | Pack of 10 |
| 11567103 | 33 mm | 38.5 mm | 38.5 mm | Pack of 5 |
| 11597103 | 39 mm | 44.5 mm | 38.5 mm | Pack of 5 |
| 11517113 | 43 mm | 49 mm | 42 mm | Pack of 2 |
| 11537113 | 49 mm | 55 mm | 42 mm | Pack of 2 |
| 11587083 | 4 mm | 5.8 mm | 14 mm | Pack of 50 |
| 11557113 | 57 mm | 64 mm | 49 mm | Each |
| 11567113 | 62 mm | 69 mm | 49 mm | Each |
| 11577113 | 67 mm | 74 mm | 49 mm | Each |
| 11507093 | 6 mm | 8 mm | 16 mm | Pack of 20 |
| 11587113 | 71 mm | 78 mm | 49 mm | Each |
| 11597113 | 75 mm | 85 mm | 54 mm | Each |
| 11517093 | 7 mm | 9 mm | 16 mm | Pack of 20 |
| 11537093 | 9 mm | 11.5 mm | 20 mm | Pack of 20 |
| 11507123 | 79 mm | 89 mm | 54 mm | Each |
| 11517123 | 86 mm | 96 mm | 54 mm | Each |
| 11527123 | 92 mm | 102 mm | 54 mm | Each |
| 11537123 | 99 mm | 109 mm | 54 mm | Each |
| 11577083 | 3 mm | 4.5 mm | 12 mm | Pack of 50 |
| 10587064 | 3 mm | 4.5 mm | 12 mm | Pack of 50 |
| 11567093 | 13 mm | 16 mm | 24 mm | Pack of 20 |
| 10265114 | 13 mm | 16 mm | 24 mm | Pack of 20 |
Specifications
Specifications
| For Use With (Application) | For laboratory use |
| Hardness | 46/55 IRHD |
| Length (Metric) | 45.5 mm |
| Quantity | 1/Ea. |
| Color | Gray |
| Diameter (Metric) Bottom | 53 mm |
| Diameter (Metric) Top | 59.5 mm |
| Material | Neoprene |
| Product Type | Bung |