Learn More
MP Biomedicals™ Sodium Free Salt Mixture for Rat, Modified
Product Code. 11487311 Shop All MP Biomedicals Products
Click to view available options
Quantity:
2 kg
10 kg
50 kg
Unit Size:
10kg
2kg
50kg
Product Code. 11487311
Brand: MP Biomedicals™ undefined
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Sodium-free salt mixture modified for rat diet.
  • Sodium Free Mineral Mixture
  • For rodent diet
  • Powder
TRUSTED_SUSTAINABILITY
Specifications
Product Type Animal Diet
For Use With (Application) Rodent diet
Quantity 50 kg
Storage Requirements Room Temperature