Learn More
MP Biomedicals™ Sodium Free Salt Mixture for Rat, Modified
Product Code. 11467311 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
2 kg
10 kg
50 kg
Unit Size:
10kg
2kg
50kg
Product Code. Quantity unitSize
11467311 2 kg 2kg
11477311 10 kg 10kg
11487311 50 kg 50kg
3 options
Product Code. 11467311
Brand: MP Biomedicals™ 0296035202
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Sodium-free salt mixture modified for rat diet.
  • Sodium Free Mineral Mixture
  • For rodent diet
  • Powder
TRUSTED_SUSTAINABILITY
Specifications
Product Type Animal Diet
For Use With (Application) Rodent diet
Quantity 2 kg
Storage Requirements Room Temperature