Learn More
Fisherbrand™ Soda Lime Glass Light Wall Rimmed Test Tube
Shop All Fisherbrand Products
Click to view available options
Diameter (Metric):
10 mm
12 mm
16 mm
24 mm
Length (Metric):
75 mm
100 mm
125 mm
150 mm
Unit Size:
Pack of 100
Pack of 50
Specifications
Specifications
| Certifications/Compliance | ISO 4142 |
| Length (Metric) | 125 mm |
| Material | Soda Lime Glass |
| Quantity | 100 Pack |
| Rimmed | Yes |
| Diameter (Metric) | 16 mm |