Learn More
Fisherbrand™ Soda Lime Glass Light Wall Rimmed Test Tube
Product Code. 15812265 Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (Metric):
10 mm
12 mm
16 mm
24 mm
Length (Metric):
75 mm
100 mm
125 mm
150 mm
Unit Size:
Pack of 100
Pack of 50
Product Code. Diameter (Metric) Length (Metric) unitSize
15812265 12 mm 75 mm Pack of 100
15892255 10 mm 75 mm Pack of 100
15832265 12 mm 100 mm Pack of 100
15852265 16 mm 125 mm Pack of 100
15872265 16 mm 150 mm Pack of 100
15842275 24 mm 150 mm Pack of 50
6 options
Product Code. 15812265
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Manufactured from soda lime glass and constructed with light walls. Fisherbrand™ Soda Lime Glass Light Wall Rimmed Test Tubes are ideal for general mixing and laboratory work.
  • Withstands moderate warming
  • Light weight
  • Complies with ISO 4142
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 4142
Length (Metric) 75 mm
Material Soda Lime Glass
Quantity 100 Pack
Rimmed Yes
Diameter (Metric) 12 mm