Learn More
Proteintech SNRPC Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22428-1-AP-150UL
Additional Details : Weight : 0.00020kg
Description
The SNRPC antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human SNRPC. This antibody recognizes human antigen. The SNRPC antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
SNRPC | |
Polyclonal | |
Unconjugated | |
150 μL | |
6631 | |
IgG |
Immunohistochemistry, ELISA | |
407 μg/mL | |
Rabbit | |
Primary | |
Human |