missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNRPC Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | SNRPC |
|---|---|
| Concentration | 407 μg/mL |
| Applications | Immunohistochemistry, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17521346
|
Proteintech
22428-1-AP-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17511346
|
Proteintech
22428-1-AP-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SNRPC antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human SNRPC. This antibody recognizes human antigen. The SNRPC antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
| SNRPC | |
| Immunohistochemistry, ELISA | |
| Unconjugated | |
| Human | |
| IgG |
| 407 μg/mL | |
| Polyclonal | |
| Rabbit | |
| 6631 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title