Learn More
SNRPC Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22428-1-AP-20UL
 View more versions of this product
Product Code. 17521346
  • 1710.33 NOK / 20µL
Description
The SNRPC antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human SNRPC. This antibody recognizes human antigen. The SNRPC antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
Specifications
SNRPC
Polyclonal
Unconjugated
20 μL
6631
IgG
Immunohistochemistry, ELISA
407 μg/mL