Learn More
Thermo Scientific Chemicals Smoothened Agonist Dihydrochloride Dihydrate
Product Code. 15416549
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Product Code. Quantity unitSize
15416549 5 mg 5mg
15426549 1 mg 1mg
2 options
Product Code. 15416549
Brand: Thermo Scientific Chemicals J64726.#0
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
A highly potent activator of Smoothened
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 364590-63-6
Molecular Formula C28H28ClN3OS.2HCl.2H2O, C28H32Cl3N3O2S
Synonym SAG1.3 HCl
Specifications
Quantity 5 mg
Description Pale yellow
Content And Storage 2°C to 8°C
For Use With (Application) A highly potent activator of Smoothened
Formula Weight 599.00
Sensitivity Light Sensitive and Hygroscopic
Product Type Dihydrochloride Dihydrate
Form Solid
InChI Key IYXUQUYRWPGIQL-UHFFFAOYSA-N
SMILES O.Cl.Cl.CNC1CCC(CC1)N(CC1=CC(=CC=C1)C1=CC=NC=C1)C(=O)C1=C(Cl)C2=CC=CC=C2S1
IUPAC Name 3-chloro-N-[4-(methylamino)cyclohexyl]-N-{[3-(pyridin-4-yl)phenyl]methyl}-1-benzothiophene-2-carboxamide hydrate dihydrochloride
Molecular Weight (g/mol) 580.99