CAS RN 364590-63-6
CAS RN 364590-63-6
IUPAC Name:
3-chloro-N-[4-(methylamino)cyclohexyl]-N-{[3-(pyridin-4-yl)phenyl]methyl}-1-benzothiophene-2-carboxamide hydrate dihydrochloride
Synonyms:
smoothened agonist hydrate dihydrochloride
Molecular Weight (g/mol):
580.99
Molecular Formula:
C28H32Cl3N3O2S
InChi Key:
IYXUQUYRWPGIQL-UHFFFAOYSA-N
SMILES:
O.Cl.Cl.CNC1CCC(CC1)N(CC1=CC(=CC=C1)C1=CC=NC=C1)C(=O)C1=C(Cl)C2=CC=CC=C2S1
1
–
1
of
1
results
1
Thermo Scientific™ Smoothened Agonist Dihydrochloride Dihydrate
CAS: 364590-63-6 Molecular Formula: C28H28ClN3OS.2HCl.2H2O,C28H32Cl3N3O2S Synonym: SAG1.3 HCl