Learn More
Miele Professional SlimLine Lab Washer PLW 7111 S-1018
Product Code. 30130362 Shop All Miele Products
Change view
Click to view available options
Volume (Metric):
241 L
Unit Size:
1 unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
30130362 241 L 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30130362 Supplier Miele Professional Supplier No. 11875200
  • ONLINE EXCLUSIVE PRICE
    1057861.50 NOK
    0.16 NOK / 1 units
    Save 1057861.34 NOK
    100% Off
 Restricted Item
Estimated Shipment: 10-11-2026
SlimLine lab washer with electric heating, hot air drying, drain pump, capacitor, water softener, conductivity meter, and oil-based app.
  • Capacity/cycle 192 narrow neck tubes or 98 pipettes - 241 L usable volume
  • Simple and flexible - EasyLoad with flexible level positions
  • Efficient - strong pump power with the variable-speed pump
  • High capacity - reprocessing of laboratory flasks up to 50 L
TRUSTED_SUSTAINABILITY
Specifications
Display Full-color Touch Screen
Material Stainless Steel
Product Type Laboratory Glassware Washer
Volume (Metric) 241 L
Color Gray
For Use With (Application) Laboratory Glassware Reprocessing
Includes Electrical Heating, Hot Air Drying, Drain Pump, Capacitor, Water Softener, Conductivity Meter, Oil-based App
Depth (English) 25 5/8 in.
Depth (Metric) 702 mm
Height (Metric) 1965 mm
Width (Metric) 650 mm
Weight (Metric) Shipping 251 kg
Electrical Requirements 3N AC 380/415 V 50 Hz
Voltage 400 V