missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC27A3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12943-1-AP-20UL
This item is not returnable.
View return policy
Description
SLC27A3 has acyl-CoA ligase activity for long-chain and very-long-chain fatty acids.SLC27A3 does not exhibit fatty acid transport activity.Specifications
| SLC27A3 | |
| Polyclonal | |
| Unconjugated | |
| SLC27A3 | |
| ACSVL3, FATP 3, FATP3, Fatty acid transport protein 3, SLC27A3, UNQ367/PRO703, VLCS 3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11000, 26568, 295219 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O88561, Q5K4L6 | |
| Slc27a3 | |
| SLC27A3 Fusion Protein Ag4015 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction