Learn More
Gilson™ Single Channel PIPETMANTM M
GREENER_CHOICE
Product Code. p-7161869 Shop All Gilson Products
Change view
Click to view available options
Volume (Metric):
0.5 to 10 μL
1 to 10 mL
100 to 1200 μL
2 to 20 μL
20 to 200 μL
20 to 300 μL
5 to 100 μL
500 to 5000 μL
Unit Size:
Each
Product Code. Volume (Metric) unitSize
16348147 0.5 to 10 μL Each
16358147 2 to 20 μL Each
16368147 5 to 100 μL Each
16378147 20 to 200 μL Each
16388147 20 to 300 μL Each
16398147 100 to 1200 μL Each
16308157 500 to 5000 μL Each
16318157 1 to 10 mL Each
8 options
Product Code. 16348147
Brand: Gilson™ F81040
Single Channel PIPETMANTM M is a BluetoothTM-enabled electronic pipette that offers accuracy and precision in both standard and repetitive pipetting modes.
  • Allows accurate and precise pipetting
  • The motorized piston allows for high pipetting reproducibility and accuracy in all modes
  • Associated with TRACKMANTM Connected and its PipettePilotTM application, PIPETMANTM M Connected reduces the risk of human error by transferring verifiable pipetting parameters and commands in the pipette
  • Increased functionality
  • The intuitive interface has five pipetting modes for a large number of applications: pipette, repetitive, mix, reverse, and custom
  • Create and transfer up to 10 custom protocols from PIPETMANTM M Connected Software into your pipette
  • BluetoothTM connection enables communication with TRACKMANTM Connected and helps guide you through your protocol telling you where, when, and how much to pipette with real-time interactions
  • Simple and comfortable to use
  • Aspirate and dispense with one click of the push button and eject tips with ease
  • The lightweight and ergonomic design rests comfortably in your hand for maximum pipetting comfort
TRUSTED_SUSTAINABILITY
Specifications
Compatible Tips Universal
Adjustable Tip Spacing No
Autoclavable Autoclavable
Color Yellow
Disposable Single-use
For Use With Pipette Tip, TRACKMAN™ Connected
Material Polypropylene
Model P10M BT
Number of Channels 1
Product Line Pipetman M
Programmability Yes
Sterility Non-sterile
Type Pipette
Volume (Metric) 0.5 to 10 μL
For Use With (Equipment) Pipette Tips, TRACKMAN™ Connected
Quantity 1
Warranty 1 Year
Length (English) 12.75 in.
Length (Metric) 32.4 cm
Width (English) 8 in.
Height (English) 2.5 in.
Height (Metric) 6.4 cm
Electrical Requirements 3.7 V and 420 mAh (1.55 Wh)
Fit for Purpose Certified Quality