Learn More
Fisherbrand™ Silicone Tubing
Product Code. 10664372 Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (Metric) Inner:
1.0 mm
2 mm
3 mm
4 mm
5 mm
6 mm
6.5 mm
8 mm
9 mm
10.0 mm
12.5 mm
16 mm
Thickness (Metric):
0.75 mm
1.0 mm
1.5 mm
2 mm
2.0 mm
2.25 mm
25 mm
Unit Size:
10m
Product Code. Diameter (Metric) Inner Thickness (Metric) unitSize
10664372 10.0 mm 2.0 mm 10m
10430313 1.0 mm 1.0 mm 10m
10726931 12.5 mm 2.25 mm 10m
10727121 16 mm 25 mm 10m
10101801 2 mm 1.0 mm 10m
10787121 3 mm 0.75 mm 10m
10111801 3 mm 1.0 mm 10m
10644182 4 mm 1.5 mm 10m
10539201 5 mm 1.5 mm 10m
11507083 6 mm 1.5 mm 10m
10549201 6.5 mm 1.5 mm 10m
11527083 8 mm 2 mm 10m
10101701 9 mm 2 mm 10m
13 options
Product Code. 10664372
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For consistently reliable and accurate fluid transfers.
Available in different diameters
Specifications
Diameter (Metric) Inner 10.0 mm
Overall Length (Metric) 10 m
Thickness (Metric) 2.0 mm