Learn More
Fisherbrand™ Silicone Tubing
Product Code. 10539201 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Diameter (Metric) Inner:
1.0 mm
2 mm
3 mm
4 mm
5 mm
6 mm
6.5 mm
8 mm
9 mm
10.0 mm
12.5 mm
16 mm
Thickness (Metric):
0.75 mm
1.0 mm
1.5 mm
2 mm
2.0 mm
2.25 mm
25 mm
Unit Size:
10m
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Thickness (Metric) Unit Size
10539201 5 mm 1.5 mm 10m
10430313 1.0 mm 1.0 mm 10m
10664372 10.0 mm 2.0 mm 10m
10726931 12.5 mm 2.25 mm 10m
10727121 16 mm 25 mm 10m
10101801 2 mm 1.0 mm 10m
10787121 3 mm 0.75 mm 10m
10111801 3 mm 1.0 mm 10m
10644182 4 mm 1.5 mm 10m
11507083 6 mm 1.5 mm 10m
10549201 6.5 mm 1.5 mm 10m
11527083 8 mm 2 mm 10m
10101701 9 mm 2 mm 10m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 10539201
  • ONLINE EXCLUSIVE PRICE
    437.98 NOK
    320.33 NOK / 10m
    Save 117.65 NOK
    26% Off
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
For consistently reliable and accurate fluid transfers.
Available in different diameters
Specifications
Diameter (Metric) Inner 5 mm
Overall Length (Metric) 10 m
Thickness (Metric) 1.5 mm