Learn More
Fisherbrand™ Silicone Stoppers
Product Code. p-8013169 Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (Metric) Bottom:
3 mm
6 mm
8 mm
13 mm
17 mm
31 mm
43 mm
49 mm
53 mm
Quantity:
1/Pk.
10/Pk.
2/Pk.
20/Pk.
50/Pk.
Unit Size:
Each
Pack of 10
Pack of 2
Pack of 20
Pack of 50
Product Code. Diameter (Metric) Bottom Quantity unitSize
11527662 13 mm 10/Pk. Pack of 10
10131511 13 mm 10/Pk. Pack of 10
11547662 17 mm 10/Pk. Pack of 10
10040131 17 mm 10/Pk. Pack of 10
10060141 31 mm 2/Pk. Pack of 2
11517672 31 mm 2/Pk. Pack of 2
10283601 3 mm 50/Pk. Pack of 50
11537652 3 mm 50/Pk. Pack of 50
11577672 43 mm 1/Pk. Each
10181841 43 mm 1/Pk. Each
11597672 49 mm 1/Pk. Each
10599561 49 mm 1/Pk. Each
11507682 53 mm 1/Pk. Each
10440693 53 mm 1/Pk. Each
11567652 6 mm 50/Pk. Pack of 50
10757311 6 mm 50/Pk. Pack of 50
11587652 8 mm 20/Pk. Pack of 20
10213751 8 mm 20/Pk. Pack of 20
18 options
Product Code. 11527662
Only null left
Add to basket
Add to basket
For laboratory and/or food testing, industrial masking or educational use.
  • Non-toxic, translucent
  • Temperature range -60°C to 200°C
  • High electrical resistance
  • Resists cracking and spallation
  • High resistance to many chemicals including ozone
Specifications
Quantity 10/Pk.
Diameter (Metric) Bottom 13 mm
Material Silicone
Product Type Stopper