Description
- Silicone
Specifications
Specifications
| Autoclavable | Autoclavable |
| Diameter (Metric) Inner | 2.8 mm |
| Material | Silicone |
| Diameter (Metric) Outer | 5 mm |
| Overall Length (Metric) | 300 cm |
| For Use With | HF2 High flow pump head |
| Sterility | Sterile |
| Flow Rate | 31mL/min. |
| Gas Permeability | High |
| Color | Pink/White |
| Show More |