Description
- Tube has no smell, taint or reaction with chlorine
- Platinum cured tubing has higher clarity with no yellowing, as experienced with peroxide cured materials
- 60°IRHD ±5°C
- FDA, BGA and WRC approved
- Temperature range: -60°C to 200°C
- Anti-adherent, hydrophobic
- High electrical and ozone resistance
- Less bacteriological build up
- Can be used for food, laboratory, pharmaceutical and medical applications
Specifications
Specifications
| Certifications/Compliance | FDA, BGA and WRC approved |
| Material | Platinum-cured Silicone |
| Diameter (Metric) Outer | 3 mm |
| Thickness (Metric) | 1 mm |
| For Use With (Application) | For food, laboratory, pharmaceutical and medical applications |
| Color | Translucent |